Skip to Content
Merck
All Photos(1)

Key Documents

N3266

Sigma-Aldrich

Neuropeptide Y porcine

≥97% (HPLC), synthetic

Synonym(s):

NPY

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C190H287N55O57
CAS Number:
Molecular Weight:
4253.65
MDL number:
UNSPSC Code:
12352200

Pricing and availability is not currently available.

biological source

synthetic

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

InChI

1S/C190H286N54O58/c1-16-94(9)149(180(296)234-129(81-140(193)253)169(285)226-124(74-93(7)8)172(288)239-150(95(10)17-2)181(297)240-151(100(15)247)182(298)221-116(32-23-67-208-190(202)203)156(272)220-119(59-62-145(260)261)161(277)218-114(30-21-65-206-188(198)199)157(273)223-121(152(195)268)76-102-39-49-108(249)50-40-102)238-173(289)127(79-105-45-55-111(252)56-46-105)229-168(284)128(80-106-86-204-90-210-106)230-159(275)115(31-22-66-207-189(200)201)219-165(281)123(73-92(5)6)224-155(271)97(12)212-174(290)134(88-245)236-167(283)126(78-104-43-53-110(251)54-44-104)228-166(282)125(77-103-41-51-109(250)52-42-103)227-158(274)113(29-20-64-205-187(196)197)216-153(269)96(11)211-163(279)122(72-91(3)4)225-170(286)131(84-147(264)265)232-162(278)118(58-61-144(258)259)217-154(270)98(13)213-177(293)137-34-25-68-241(137)183(299)99(14)214-164(280)130(83-146(262)263)231-160(276)117(57-60-143(256)257)215-142(255)87-209-176(292)136-33-24-70-243(136)186(302)133(82-141(194)254)235-171(287)132(85-148(266)267)233-178(294)139-36-27-71-244(139)185(301)120(28-18-19-63-191)222-175(291)135(89-246)237-179(295)138-35-26-69-242(138)184(300)112(192)75-101-37-47-107(248)48-38-101/h37-56,86,90-100,112-139,149-151,245-252H,16-36,57-85,87-89,191-192H2,1-15H3,(H2,193,253)(H2,194,254)(H2,195,268)(H,204,210)(H,209,292)(H,211,279)(H,212,290)(H,213,293)(H,214,280)(H,215,255)(H,216,269)(H,217,270)(H,218,277)(H,219,281)(H,220,272)(H,221,298)(H,222,291)(H,223,273)(H,224,271)(H,225,286)(H,226,285)(H,227,274)(H,228,282)(H,229,284)(H,230,275)(H,231,276)(H,232,278)(H,233,294)(H,234,296)(H,235,287)(H,236,283)(H,237,295)(H,238,289)(H,239,288)(H,240,297)(H,256,257)(H,258,259)(H,260,261)(H,262,263)(H,264,265)(H,266,267)(H4,196,197,205)(H4,198,199,206)(H4,200,201,207)(H4,202,203,208)/t94-,95-,96-,97-,98-,99-,100+,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,126-,127-,128-,129-,130-,131-,132-,133-,134-,135-,136-,137-,138-,139-,149-,150-,151-/m0/s1

InChI key

JYLKGTCHMPLCES-OFGSCBOVSA-N

Gene Information

pig ... NPY(397304)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2

Biochem/physiol Actions

Vasoconstrictor; brain peptide that inhibits Ca2+-activated K+ channels in vascular smooth muscle. Implicated in the control of blood pressure, sexual behavior and food intake. Inhibits cholecystokinin- and secretin-stimulated pancreatic secretion.
Vasoconstrictor; inhibits Ca2+-activated K+ channels in vascular smooth muscle; inhibits cholecystokinin- and secretin-stimulated pancreatic secretion.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kavita N Hadawale et al.
Neuropeptides, 74, 1-10 (2019-03-04)
Neuropeptide Y (NPY) is involved in sex-specific behavioural processes in vertebrates. NPY integrates energy balance and reproduction in mammals. However, the relevance of NPY in reproduction of lower vertebrates is understudied. In the present study, we have investigated neuroanatomical distribution
T Onaga et al.
Domestic animal endocrinology, 74, 106523-106523 (2020-08-17)
The present study aimed to determine the primary sequence of ovine xenin and clarify the mRNA expression and peptide localization of xenin in the gastrointestinal tract in sheep. The colocalization of xenin and glucose-dependent insulinotropic polypeptide was also compared in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service