E5627
Erythropoietin human
recombinant, expressed in CHO cells, lyophilized powder, suitable for cell culture, ~100,000 units/mg protein
Synonym(s):
EPO
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in CHO cells
form
lyophilized powder
specific activity
~100,000 units/mg protein
packaging
pkg of 10UN
technique(s)
cell culture | mammalian: suitable
color
white
UniProt accession no.
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Physical form
Analysis Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service