Q3376
Quoirin and Lepoivre Basal Salt Mixture
suitable for plant cell culture, powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
Recommended Products
form
powder
technique(s)
cell culture | plant: suitable
application(s)
agriculture
storage temp.
2-8°C
Formula variant
With the macro- and micronutrients as described by Quoirin, et al. (1977).
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service