Skip to Content
Merck
All Photos(1)

Key Documents

M7522

Sigma-Aldrich

2-Mercaptoethanol

liquid, BioReagent, suitable for cell culture

Synonym(s):

β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Beilstein:
773648
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

2.69 (vs air)

vapor pressure

1 mmHg ( 20 °C)

product line

BioReagent

form

liquid

expl. lim.

18 %

concentration

14.3 M (pure liquid)

technique(s)

cell culture | mammalian: suitable

refractive index

n20/D 1.500 (lit.)

bp

157 °C (lit.)

solubility

H2O: 1 mL/mL

density

1.114 g/mL at 25 °C (lit.)

SMILES string

OCCS

InChI

1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2

InChI key

DGVVWUTYPXICAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BME is suitable for reducing protein disulfide bonds prior to polyacrylamide gel electrophoresis and is usually included in a sample buffer for SDS-PAGE at a concentration of 5%. Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on SDS-PAGE. Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 2 Oral

Target Organs

Liver,Heart

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karl W Boehme et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(47), 19986-19991 (2009-11-10)
Serotype-specific patterns of reovirus disease in the CNS of newborn mice segregate with the viral S1 gene segment, which encodes attachment protein sigma1 and nonstructural protein sigma1s. The importance of receptor recognition in target cell selection by reovirus implicates the
Vesna Nikolova-Krstevski et al.
BMC genomics, 9, 240-240 (2008-05-24)
Endothelial differentiation occurs during normal vascular development in the developing embryo. This process is recapitulated in the adult when endothelial progenitor cells are generated in the bone marrow and can contribute to vascular repair or angiogenesis at sites of vascular
Maria Bettini et al.
Journal of neuroimmunology, 213(1-2), 60-68 (2009-06-23)
XIncreasing evidence supports a role for CD8+ T cells in multiple sclerosis. In an attempt to isolate the contribution of CD8+ T cells in a murine model of MS, we immunized mice with a dominant CD8 epitope MOG37-46, a truncated
Manching Ku et al.
PLoS genetics, 4(10), e1000242-e1000242 (2008-11-01)
In embryonic stem (ES) cells, bivalent chromatin domains with overlapping repressive (H3 lysine 27 tri-methylation) and activating (H3 lysine 4 tri-methylation) histone modifications mark the promoters of more than 2,000 genes. To gain insight into the structure and function of
Eliana B Marengo et al.
PloS one, 3(8), e3025-e3025 (2008-08-22)
The heat shock protein [Hsp] family guides several steps during protein synthesis, are abundant in prokaryotic and eukaryotic cells, and are highly conserved during evolution. The Hsp60 family is involved in assembly and transport of proteins, and is expressed at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service