Skip to Content
Merck
All Photos(2)

Key Documents

E6783

Sigma-Aldrich

pFLAG-CMV-3 Expression Vector

shuttle vector for transient or stable expression of N-terminal FLAG

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352200

tag

FLAG® tagged

grade

Molecular Biology
for molecular biology

form

buffered aqueous solution

Peptide tag location

N-terminal

shipped in

dry ice

storage temp.

−20°C

General description

The pFLAG-CMV-3 Expression Vector is a 6.3 kb vector used for transient or stable expression in
mammalian cells. The vector is a derivative of pCMV5 used for transient or stable expression and secretion of a properly inserted open reading frame as an N-terminal FLAG fusion protein.

The pFLAG-CMV-3 Expression Vector is a shuttle vector containing both bacterial and SV40 origins of replication for propagation in both E. coli and mammalian cells. Efficiency of replication and genomic integration is optimal when using host cells that express the SV40 large T antigen.

The pFLAG-CMV-3-BAP Control Plasmid is a 7.7 kb derivative of pCMV51 used for transient expression and secretion of N-terminal FLAG bacterial alkaline phosphatase fusion protein in mammalian cells.

The FLAG® epitope is a small, hydrophilic 8 amino acid tag (DYKDDDDK) that provides for sensitive detection and high quality purification using ANTI-FLAG products available from Sigma-Aldrich.

Vector Maps and Sequences

Application

pFLAG-CMV-3 Expression Vector is suitable for transient or stable expression and secretion of N-terminal FLAG-tagged fusion proteins in mammalian cells.

Components

  • pFLAG-CMV-3 Expression Vector 20 μg (E8770) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • pFLAG-CMV-3-BAP Control Plasmid 20 μg (C3972) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

The promoter-regulatory region of the human cytomegalovirus drives transcription of FLAG-fusion constructs. The preprotrypsin leader sequence precedes the FLAG® sequence and directs secretion of the fusion protein into the culture medium. The aminoglycoside phosphotransferase II gene (neo-r) confers resistance to aminoglycosides such as G 418, allowing for selection of stable transfectants.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koichi Ushizawa et al.
PloS one, 4(6), e5814-e5814 (2009-06-09)
Ly-6 superfamily members have a conserved Ly-6 domain that is defined by a distinct disulfide bonding pattern between eight or ten cysteine residues. These members are divided into membrane-type and secretory-type proteins. In the present study, we report the identification
Aidan L Huene et al.
iScience, 24(7), 102811-102811 (2021-07-24)
Many organisms use genetic self-recognition systems to distinguish themselves from conspecifics. In the cnidarian, Hydractinia symbiolongicarpus, self-recognition is partially controlled by allorecognition 2 (Alr2). Alr2 encodes a highly polymorphic transmembrane protein that discriminates self from nonself by binding in trans
M Kokel et al.
Genes & development, 12(10), 1425-1437 (1998-05-29)
Receptor tyrosine phosphatases have been implicated in playing important roles in cell signaling events by their ability to regulate the level of protein tyrosine phosphorylation. Although the catalytic activity of their phosphatase domains has been well established, the biological roles
Chenyun Wang et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 32(2), 299-302 (2012-04-25)
Many researchers employed mammalian expression system to artificially express cannabinoid receptors, but immunoblot data that directly prove efficient protein expression can hardly be seen in related research reports. In present study, we demonstrated cannabinoid receptor protein was not able to
Ahmet Sinan Sari et al.
Turkish journal of biology = Turk biyoloji dergisi, 45(3), 301-313 (2021-08-12)
Selective targeting of transfected mesenchymal stem cells (MSCs) carrying specific antioncogenes to the tumor was suggested as a treatment option. Bone morphogenetic protein-2 (BMP2) was shown to inhibit the proliferation and aggressiveness of osteosarcoma (OS) cells. Here, we aimed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service