Skip to Content
Merck

1.20078

Millipore

Eshmuno® S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511

ligand

(Sulfoisobutyl)

sterility

sterile (Caustic Stable)

product line

Eshmuno®

form

resin

specific analyte(s)

MAbs

parameter

1000 cm/hr flow rate
8 bar max. pressure

matrix active group

polymer, Polyvinyl Ether

mean particle size

85 μm

capacity

165 mg binding capacity (lysozyme/ml of resin)

transition temp

flash point 35 °C (own results)

application(s)

recombinant protein
vaccine development

separation technique

cation exchange

storage temp.

2-30°C

General description

The first member of the Eshmuno® resin family, designed specifically for highly productive downstream bioprocessing

Features and Benefits

Eshmuno® S chromatography resin enables:

  • Purification of basic and neutral proteins (e.g., antibodies) and peptides specifically designed for highly productive downstream bioprocessing
  • High flow rate applications
  • High separation efficiency over a broad range of flow rates with a rigid matrix and optimized pore structure
  • Tentacle technology which provides high yield, selectivity, and focused elution
  • Multiple CIP and packing cycles with the chemically and physically robust bead, leading to a longer resin lifetime
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Packaging

  • 1.20078.0100: Eshmuno® S Resin 100ml
  • 1.20078.0010: Eshmuno® S Resin 10ml
  • 1.20078.0500: Eshmuno® S Resin 500ml
  • 1.20078.5000: Eshmuno® S Resin 5L

Plastic bottle

Analysis Note

Appearance: Milky cream coloured turbid suspension, free from coarse mechanical impurities.
Microscopic evaluation: Spherical particles, no agglomerates, no fines.
Extractable matter (water): ≤ 0.08 %
Ionic capacity: 50 - 100 µeq/ml
Performance test (chymotrypsinogen): 10 - 30 mS/cm
Performance test (lysozyme): 35 - 55 mS/cm
Pressure drop(column: ID=1.6 cm, L=10 cm at 5 ml/min): ≤ 1.0 bar
Protein binding capacity (lysozyme): 120 - 180 mg/ml
Particle size (50-120 µm): ≥ 80 %
Particle size (d50): 75 - 95 µm
Cerium: ≤ 30 µg/g
Endotoxins: ≤ 1.00 EU/ml
TAMC (total aerobic microbial count): ≤ 100 CFU/ml
TYMC (total yeast and mould count): ≤ 100 CFU/ml

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service