Skip to Content
Merck
All Photos(1)

Key Documents

CHR100

ChIC/CUT&RUN Kit (pA-MN)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41116158

Application

A novel alternative to ChIP with a much faster procedure,extremely low signal-to-noise ratio and much less starting sample.

Components

Concanavalin A-coated magnetic beads Binding BufferWash Buffer Spermidine0.5M EDTARinse Buffer Incubation buffer Stop Solution2% Digitonin containing DMSORNase A Spike-in DNA Precipitate Enhancer Proteinase K SolutionProtease Inhibitor Cocktail III, Animal Free **Contains DMSO Rabbit IgG Purified Anti-H3K27me3 rabbit monoclonal antibodyProtein A–micrococcal nuclease (pA-MN) fusion protein

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service