Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

80489

Millipore

β-Lactamase Strips Kit

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Moraxella spp.
Enterococcus spp.
Haemophilus spp. (species)
Neisseria spp.
Staphylococcus spp.

Diagnostic Implications

β-Lactamase Strips are test strips for the rapid acidimetric detection of the β-lactamase activity of microorganisms. The test is based on hydrolysis of the β-lactam ring in benzylpenicillin, which results in the production of penicilloic acid. This process causes acidification of the bacterial suspension, and changes the colour of the acidobasic indicator. The acidobasic test for β-lactamase activity is suitable only for the detection of Haemophilus influenzae, Neisseria gonorrhoeae and Staphylococcus spp. This test is not suitable for the detection of the β-lactamase activity of other microorganisms (Branhamella catarrhalis, Enteroccocus faecalis, Neisseria meningitis, Enterococcus spp. and others). The result of the reaction is read after 2-10 min.

Components

The kit contains:
Plastic strips with an active zone saturated with benzylpenicillin and an acidobasic indicator (Cat. No. 56348) 100 pcs
Moistening solution (Cat. No.23134) 70mL

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

80489-100EA:
80489-100EA-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Linder et al.
The EMBO journal, 39(19), e105071-e105071 (2020-08-26)
Inflammasomes execute a unique type of cell death known as pyroptosis. Mostly characterized in myeloid cells, caspase-1 activation downstream of an inflammasome sensor results in the cleavage and activation of gasdermin D (GSDMD), which then forms a lytic pore in

Articles

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service