コンテンツへスキップ
Merck
Corticosteroid regulation of P-glycoprotein in the developing blood-brain barrier.
Endocrinology (2011-01-18)
Majid Iqbal, William Gibb, Stephen G Matthews
要旨
The early fetal brain is susceptible to teratogens in the maternal circulation, because brain microvessel expression of drug efflux transporter, P-glycoprotein (P-gp), is very low. However, there is a dramatic up-regulation of brain microvessel P-gp in late gestation. This study investigated the role of cortisol and dexamethasone in this up-regulation of fetal brain microvessel P-gp expression. Primary brain endothelial cell (BEC) cultures derived from gestational d (GD)40, GD50, GD65 (term, ∼68 d) and postnatal d 14 male guinea pigs were treated with varying doses (10(-8) to 10(-5) m) of cortisol, dexamethasone, and aldosterone. After treatment, P-gp function was assessed using calcein-acetoxymethyl ester (P-gp substrate; 1 μm for 1 h) and measuring BEC accumulation of calcein. Corticosteroid treatment of BECs derived from postnatal d 14 resulted in increased P-gp activity. BECs derived from GD65 (near term) responded similarly, but these cells were extremely sensitive to the effects of mineralocorticoid receptor agonists (cortisol and aldosterone). BECs derived from GD50 displayed dose-dependent increases in P-gp function with dexamethasone (P < 0.05) and a trend towards increased function with cortisol. Cells derived from GD40 were unresponsive to all treatments. In conclusion, P-gp function in BECs is more responsive to glucocorticoids (GCs) in late gestation. Therefore, the late gestational surge in fetal plasma GCs, which parallels the increase in brain microvessel P-gp expression, may contribute to this P-gp up-regulation. Further, synthetic GCs (administered to pregnant women at risk of preterm delivery) may increase the protective capacity of the developing fetal blood-brain barrier, depending on the timing of GC exposure.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
モノクロナール抗シナプトフィジン マウス宿主抗体, clone SVP-38, ascites fluid
Sigma-Aldrich
Anti-GLUT1 (CT) Antibody, clone 5B12.3, clone 5B12.3, from mouse