コンテンツへスキップ
Merck
  • Click-modified anandamide siRNA enables delivery and gene silencing in neuronal and immune cells.
Click-modified anandamide siRNA enables delivery and gene silencing in neuronal and immune cells.
Journal of the American Chemical Society (2012-07-21)
Julian Willibald, Johannes Harder, Konstantin Sparrer, Karl-Klaus Conzelmann, Thomas Carell
要旨
Click chemistry of alkyne-modified RNA with different receptor ligand azides was used to prepare 3'-folate, 3'-cholesterol, and, as a new entity, 3'-anandamide-modified RNA in high yields and excellent purity. The anandamide-modified RNA shows surprisingly high transfection properties and enables the delivery of siRNA even into difficult-to-transfect RBL-2H3 cells which model neuronal uptake. Furthermore, the system was employed in human immune cells (BJAB), demonstrating silencing effects similar to those of a cationic, benchmark transfection reagent. In addition, the anandamide conjugates were found to be nontoxic. The reported chemistry and the described properties of the anandamide siRNA extend the possibilities of using siRNA-based gene silencing in neuronal and immune cells.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
Cholesteryl-TEG azide