コンテンツへスキップ
Merck
Lipophilic organic contaminants in the Rhine river, Germany.
Water research (2005-11-11)
Jan Schwarzbauer, Sabine Heim
要旨
Detailed gas chromatographic-mass spectrometric analyses applied to eight Rhine river water samples constituted a comprehensive characterization of the low molecular weight organic contamination. Within the group of predominant anthropogenic contaminants, only a few compounds were characterized as frequently detected or priority pollutants. Numerous compounds exhibiting physiological or ecotoxicological properties are only rarely reported or still unnoticed riverine contaminants. Information on environmental behaviour or ecotoxicological effects is still limited for most of these substances. In particular, several brominated compounds (mono- and dibrominated (methoxyphenyl)propionic acids and hydroxymethylacetophenones) were identified for the first time as environmental contaminants. Quantitative analyses differentiated five groups of pollutants with respect to their concentration profiles. The spatial distribution and the intensity of emission sources on the one hand and the environmental stability as well as the tendency to adsorb on the particulate matter on the other hand determined the quantitative occurrence of individual compounds.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
フェニルプロピオル酸, 99%