コンテンツへスキップ
Merck
  • Evaluation of nitazoxanide and oxfendazole efficacy against cystic echinococcosis in naturally infected sheep.
Evaluation of nitazoxanide and oxfendazole efficacy against cystic echinococcosis in naturally infected sheep.
The American journal of tropical medicine and hygiene (2009-03-10)
Cesar M Gavidia, Armando E Gonzalez, Luis Lopera, Cesar Jayashi, Roxana Angelats, Eduardo A Barron, Berenice Ninaquispe, Lucia Villarreal, Hector H Garcia, Manuela R Verastegui, Robert H Gilman
要旨
Cystic echinococosis (CE) is a public health problem caused by Echinococcus granulosus. We aimed to determine the efficacy of nitazoxanide (NTZ) and oxfendazole (OXF) against CE in naturally infected sheep. A total of 151 ewes were assigned to the following groups: 15 mg/kg of NTZ weekly for five weeks (NTZ5); two rounds of 15 mg/ kg of NTZ a day for five days (NTZ5x2) two weeks apart; 30 mg/kg of OXF a week for 11 weeks (OXF11); 30 mg/kg of OXF plus 15 mg/kg of NTZ a week for 11 weeks (OXF/NTZ); and the control group. OXF11 and OXF/NTZ decreased the number of fertile cysts, increased the number of degenerated cysts, and were more efficacious (49.6-61.2%) against lung cysts and liver cysts (91.8-100%) than any other treatment group. OXF might be an additional strategy for control programs and an optional treatment of human CE after it is licensed.
材料
製品番号
ブランド
製品内容
Supelco
オクスフェンダゾール, VETRANAL®, analytical standard