コンテンツへスキップ
Merck
A new cytotoxic cholestane bisdesmoside from Ornithogalum saundersiae bulbs.
Bioscience, biotechnology, and biochemistry (1996-06-01)
Y Mimaki, M Kuroda, A Kameyama, Y Sashida, T Hirano, K Oka, K Koike, T Nikaido
要旨
Bioassay-guided fractionation of the MeOH extract of Ornithogalum saundersiae bulbs led to the isolation of a new cholestane bisdesmoside with potent cytotoxic activities toward leukemia HL-60 and MOLT-4 cells. The structure was deduced mainly from spectroscopic information.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
3,4,5-トリメトキシ安息香酸, ReagentPlus®, 99%