コンテンツへスキップ
Merck
Role of postreplicative DNA mismatch repair in the cytotoxic action of thioguanine.
Science (New York, N.Y.) (1996-08-23)
P F Swann, T R Waters, D C Moulton, Y Z Xu, Q Zheng, M Edwards, R Mace
要旨
It is proposed here that the delayed cytotoxicity of thioguanine involves the postreplicative DNA mismatch repair system. After incorporation into DNA, the thioguanine is chemically methylated by S-adenosylmethionine to form S6-methylthioguanine. During DNA replication, the S6-methylthioguanine directs incorporation of either thymine or cytosine into the growing DNA strand, and the resultant S6-methylthioguanine-thymine pairs are recognized by the postreplicative mismatch repair system. Azathioprine, an immunosuppressant used in organ transplantation, is partly converted to thioguanine. Because the carcinogenicity of N-nitrosamines depends on formation of O6-alkylguanine in DNA, the formation of the analog S6-methylthioguanine during azathioprine treatment may partly explain the high incidence of cancer after transplantation.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
2-Amino-6-methylmercaptopurine, ≥95%