コンテンツへスキップ
Merck
Alteration of serum N-glycan profile in patients with autoimmune pancreatitis.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.] (2016-01-03)
Takeshi Tomoda, Kazuhiro Nouso, Hironari Kato, Koji Miyahara, Chihiro Dohi, Yuki Morimoto, Hideaki Kinugasa, Yutaka Akimoto, Kazuyuki Matsumoto, Naoki Yamamoto, Yasuhiro Noma, Shigeru Horiguchi, Koichiro Tsutsumi, Maho Amano, Shin-Ichiro Nishimura, Kazuhide Yamamoto
要旨
The aims of this study were to determine the change in whole-serum N-glycan profile in autoimmune pancreatitis (AIP) patients and to investigate its clinical utility. We collected serum from 21 AIP patients before any treatment, and from 60 healthy volunteers (HLTs). Serum glycan profile was measured by comprehensive and quantitative high-throughput glycome analysis. Of the 53 glycans detected, 14 were differentially expressed in AIP patients. Pathway analysis demonstrated that agalactosyl and monogalactosyl bi-antennary glycans were elevated in AIP patients. Among the 14 glycans, #3410, #3510, and #4510 showed high area under receiver operating characteristic (AUROC) values (0.955, 0.964, and 0.968 respectively) for the diagnosis of AIP. These three glycans were mainly bound to immunoglobulin G; however, their serum levels were significantly higher, even in AIP patients who showed lower serum IgG4 levels, than in HLTs. We demonstrated, for the first time, whole-serum glycan profiles of AIP patients and showed that the levels of glycans #3410, #3510, and #4510 were increased in AIP patients. These glycans might be valuable biomarkers of AIP.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
O-ベンジルヒドロキシルアミン, AldrichCPR