コンテンツへスキップ
Merck
  • Prolactin and glucocorticoid signaling induces lactation-specific tight junctions concurrent with β-casein expression in mammary epithelial cells.
Prolactin and glucocorticoid signaling induces lactation-specific tight junctions concurrent with β-casein expression in mammary epithelial cells.
Biochimica et biophysica acta (2016-05-01)
Ken Kobayashi, Yusaku Tsugami, Kota Matsunaga, Shoko Oyama, Chinatsu Kuki, Haruto Kumura
要旨
Alveolar mammary epithelial cells (MECs) in mammary glands are highly specialized cells that produce milk for suckling infants. Alveolar MECs also form less permeable tight junctions (TJs) to prevent the leakage of milk components after parturition. In the formation process of less permeable TJs, MECs show a selective downregulation of Cldn4 and a localization change of Cldn3. To investigate what induces less permeable TJs through these compositional changes in Cldns, we focused on two lactogenesis-related hormones: prolactin (Prl) and glucocorticoids. Prl caused a downregulation of Cldn3 and Cldn4 with the formation of leaky TJs in MECs in vitro. Prl-treated MECs also showed low β-casein expression with the activation of STAT5 signaling. By contrast, dexamethasone (Dex), a glucocorticoid analogue, upregulated Cldn3 and Cldn4, concurrent with the formation of less permeable TJs and the activation of glucocorticoid signaling without the expression of β-casein. Cotreatment with Prl and Dex induced the selective downregulation of Cldn4 and the concentration of Cldn3 in the region of TJs concurrent with less permeable TJ formation and high β-casein expression. The inhibition of Prl secretion by bromocriptine in lactating mice induced the upregulation of Cldn3 and Cldn4 concurrent with the downregulation of milk production. These results indicate that the coactivation of Prl and glucocorticoid signaling induces lactation-specific less permeable TJs concurrent with lactogenesis.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
モノクロナール抗サイトケラチン, パン (混合物) マウス宿主抗体, clone C-11+PCK-26+CY-90+KS-1A3+M20+A53-B/A2, ascites fluid
Sigma-Aldrich
抗GR ウサギ宿主抗体, affinity isolated antibody