コンテンツへスキップ
Merck
  • ACOX2 deficiency: A disorder of bile acid synthesis with transaminase elevation, liver fibrosis, ataxia, and cognitive impairment.
ACOX2 deficiency: A disorder of bile acid synthesis with transaminase elevation, liver fibrosis, ataxia, and cognitive impairment.
Proceedings of the National Academy of Sciences of the United States of America (2016-09-21)
Sílvia Vilarinho, Sinan Sari, Francesca Mazzacuva, Kaya Bilgüvar, Güldal Esendagli-Yilmaz, Dhanpat Jain, Gülen Akyol, Buket Dalgiç, Murat Günel, Peter T Clayton, Richard P Lifton
要旨
Acyl CoA Oxidase 2 (ACOX2) encodes branched-chain acyl-CoA oxidase, a peroxisomal enzyme believed to be involved in the metabolism of branched-chain fatty acids and bile acid intermediates. Deficiency of this enzyme has not been described previously. We report an 8-y-old male with intermittently elevated transaminase levels, liver fibrosis, mild ataxia, and cognitive impairment. Exome sequencing revealed a previously unidentified homozygous premature termination mutation (p.Y69*) in ACOX2 Immunohistochemistry confirmed the absence of ACOX2 expression in the patient's liver, and biochemical analysis showed marked elevation of intermediate bile acids upstream of ACOX2. These findings define a potentially treatable inborn error of bile acid biosynthesis caused by ACOX2 deficiency.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
Anti-ACOX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution