GCによる5-アミノ-1,3,3-トリメチルシクロヘキサンメチルアミンエナンチオマー (N-TFA 誘導体) の分析、Astec® CHIRALDEX™ A-TA
CONDITIONS
column
Astec CHIRALDEX A-TA, 30 m x 0.25 mm I.D., 0.12 μm (73003AST)
oven
160 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
1 μL, 80:1 split
sample
3 mg/mL racemate in methanol
peaks 1 & 2: first enantiomer pair
peaks 3 & 4: second enantiomer pair