化粧品保存料の分析、Ascentis Express RP-Amide、温度上昇率(0.6 mL/min)
材料
分析カラム
column
Ascentis Express RP-Amide, 10 cm x 3 mm I.D., 2.7 μm particles (53918-U)
mobile phase
[A] water; [B] methanol; (60:40, A:B)
flow rate
0.6 mL/min
column temp.
50 °C
detector
UV, 250 nm
injection
1 μL
sample
methyl-, benzylparaben, 50 mg/L; ethylparaben, 30 mg/L; propyl-, butylparaben, 100 mg/L; 2-phenoxyethanol, 1.5 g/L; 1-phenoxy-2-propanol, 3 g/L; all in 40% methanol