ダイエットソーダ添加物の分析、Ascentis Express RP-Amide
材料
Standard
分析カラム
column
Ascentis Express RP-Amide, 3 cm x 4.6 mm I.D., 2.7 μm particles (53921-U)
mobile phase
[A] 20 mM ammonium acetate, pH 5.6 with acetic acid; [B] water; [C] acetonitrile
gradient
20% A constant; 75 to 20% B, 5 to 60% C in 1 min; held at 20% B, 60% C for 0.1 min
pressure
3336 psi (230 bar)
flow rate
3.0 mL/min
column temp.
40 °C
detector
UV, 214 nm
injection
1 μL
sample
Diet Soda 100 - 500 μg/mL in buffer
詳細
アナリシスノート
This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in diet cola.