HPLCによる市販の殺菌シートに含まれる塩化ベンザルコニウムの分析、Ascentis Express HILIC
材料
分析カラム
column
Ascentis Express HILIC, 15 cm x 4.6 mm I.D., 2.7 μm particles (53981-U)
mobile phase
[A] 20 mM ammonium acetate; [B] acetonitrile; (10:90, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
ESI(+), TIC 150 - 500 m/z
injection
2 μL
sample
Single wipe sheet, extracted with 10 mL methanol