HPLCによるニカルジピンエナンチオマーの分析、CHIROBIOTIC® V2
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
[A] methanol; [B] 20 mM ammonium acetate, pH 4.1; (95:5, A:B)
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 230 nm