S5791
SPIT Medium Supplement (100×)
liquid, sterile-filtered, BioReagent, suitable for cell culture
About This Item
Recommended Products
sterility
sterile-filtered
product line
BioReagent
form
liquid
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
storage temp.
2-8°C
Application
INSULIN is a polypeptide hormone that promotes the uptake of glucose and amino acids and may owe observed mitogenic effect to this property.
TRANSFERRIN is an iron-transport protein. Iron is an essential trace element, but can be toxic in the free form. To nourish cells in culture, it is supplied bound to transferrin in serum.
SELENIUM is an essential trace element normally provided by Serum.
SODIUM PYRUVATE has been shown to be beneficial as an additional energy source in some instances.
ETHANOLAMINE is a fatty acid that plays a significant role in the proliferation of hybridoma cells and frequently is added to supplements used for culturing these cells.
Nutritional studies indicate that the supplement components described are utilized by most mammalian cells. They enhance cell proliferation and decrease the serum requirement of many cell types. When the provided supplement is used with 2 to 4 percent serum, proliferation is reported to be similar to medium supplemented with 10 percent serum.
Components
Other Notes
Preparation Note
related product
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service