뒤로
Kern PLS series precision balances
Products
제품 카테고리
balances (6)
Manufacturer
칭량 용량(g)
포장 단위
제품 필터
Showing 1-6 of 6
비교
제품 번호
설명
가격
Z742869
Kern PLS series precision balances,
Kern PLS 1200-3A, weighing capacity 1200 g, accuracy: 0.001 g, precision: ±0.003 g
확장
가격 보기
Z742871
Kern PLS series precision balances,
Kern PLS 6200-2A, weighing capacity 6200 g, accuracy: 0.01 g, precision: ±0.03 g
확장
가격 보기
Z742868
Kern PLS series precision balances,
Kern PLS 720-3A, weighing capacity 720 g, accuracy: 0.001 g, precision: ±0.002 g
확장
가격 보기
Z742867
Kern PLS series precision balances,
Kern PLS 420-3F, weighing capacity 420 g, accuracy: 0.001 g, precision: ±0.004 g
확장
가격 보기
Z742873
Kern PLS series precision balances,
Kern PLS 8000-2A, weighing capacity 8000 g, accuracy: 0.01 g, precision: ±0.04 g
확장
가격 보기
Z742870
Kern PLS series precision balances,
Kern PLS 4200-2F, weighing capacity 4200 g, accuracy: 0.01 g, precision: ±0.04 g