뒤로
Minimum Essential Medium
동의어:
Minimum Essential Medium Eagle, MEM
Products
제품 카테고리
media (2)
브랜드
SAFC (7)
물리적 형태
얼스염(Earles Salts)
L-글루타민
중탄산 나트륨
품질 수준
멸균 방법
Technique
제품 필터
Showing 1-7 of 7
비교
제품 번호
설명
가격
51412C
Minimum Essential Medium,
with Earle′s Balanced Salts, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
확장
가격 보기
56416C
Minimum Essential Medium,
with Earle′s Balanced Salts, with 2.0 mM L-glutamine, with non-essential amino acids, without sodium bicarbonate, dry powder, suitable for cell culture
확장
가격 보기
51411C
Minimum Essential Medium,
with Earle′s Balanced Salts, with 2.0 mM L-glutamine, liquid, sterile-filtered, suitable for cell culture
확장
가격 보기
51417C
Minimum Essential Medium,
with Earle′s salts, with 2.0 mM L-glutamine, with 20.0 mM HEPES, with non-essential amino acids, liquid, sterile-filtered, suitable for cell culture
확장
가격 보기
51416C
Minimum Essential Medium,
with Earle′s Balanced Salts, with non-essential amino acids, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
확장
가격 보기
51415C
Minimum Essential Medium,
with Earle′s Balanced Salts, with 25mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
확장
가격 보기
56419C
Minimum Essential Medium,
with Earle′s Balanced Salts, with 2.0 mM L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture