뒤로
Valco
®
Precision Sampling Syringe, Series A, Removable needle
Products
제품 카테고리
syringes (2)
Manufacturer
길이(in)
Volume (µL)
니들 모양
포장 단위
제품 필터
Showing 1-2 of 2
비교
제품 번호
설명
가격
22263
Valco
®
Precision Sampling Syringe, Series A, Removable needle,
volume 500 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2.25 in. × 0.029 in. × 0.012 in.
확장
가격 보기
22264
Valco
®
Precision Sampling Syringe, Series A, Removable needle,
volume 1000 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2.25 in. × 0.029 in. × 0.012 in.