콘텐츠로 건너뛰기
Merck
  • Residue behavior and risk assessment of thifluzamide in the maize field ecosystem.
Residue behavior and risk assessment of thifluzamide in the maize field ecosystem.
Environmental science and pollution research international (2018-05-20)
Cong Ma, Zhongdong Liu, Yan Qi, Shanshan Wang, Xiaolin Cao, Jing Wang, Yongxin She, Yong Shao, Jitai Shen, Chao Zhang, Zejun Jiang
초록
In the present work, the dissipation kinetics and final residue levels of thifluzamide in the maize field ecosystem were investigated. Using a modified quick, easy, cheap, effective, rugged, and safe (QuEChERS) extraction combined with liquid chromatography-tandem mass spectrometric detection (LC-MS/MS), a rapid, sensitive, efficient, and reliable method for extraction and quantitative analysis of thifluzamide residues in maize grain, maize plant, and soil was developed. Satisfactory recoveries of 78.7-97.0% were achieved with relative standard deviations (RSDs) in the range of 1.6 to 8.2%. The limits of detection (LODs) and the limit of quantification (LOQ) were 0.002-0.005 and 0.010 mg kg-1, respectively. The dissipation kinetics of thifluzamide in maize plant was well fitted by the first-order kinetic model with short half-lives of 0.19-0.22 days, while thifluzamide degraded slowly in soil with half-lives of 4.56-15.85 days. The final residues in maize grain, maize plant, and soil samples collected at the milk stage and the physiological maturity stage were no more than 0.010, 0.807, and 0.278 mg kg-1, respectively. Given that no maximum residue limit (MRL) for thifluzamide in maize has been established, the safety of this fungicide application was estimated by a dietary risk assessment. The hazard quotient was 0.03%, which was substantially less than 1, indicating that the long-term risk induced by the thifluzamide application on maize at the recommended dose is negligible. These results help governments to develop regulations for the safe use of thifluzamide.
MATERIALS
제품 번호
브랜드
제품 설명
Supelco
Thifluzamide, PESTANAL®, analytical standard