콘텐츠로 건너뛰기
Merck
  • Determination of carnitine, its short chain acyl esters and metabolic precursors trimethyllysine and gamma-butyrobetaine by quasi-solid phase extraction and MS/MS detection.
Determination of carnitine, its short chain acyl esters and metabolic precursors trimethyllysine and gamma-butyrobetaine by quasi-solid phase extraction and MS/MS detection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-06-16)
Frank Hirche, Maren Fischer, Janine Keller, Klaus Eder
초록
For the investigation of the metabolism and biosynthesis of carnitine, sensitive determination of carnitine and its metabolic precursors, trimethyllysine and gamma-butyrobetaine, is required. We present here a new simplified method for the analysis of carnitine, its acetyl- and propyl esters, as well as trimethyllysine and gamma-butyrobetaine without need for derivatization reactions by means of normal-phase LC and electrospray ionization tandem mass spectrometry. The limits of quantification were between 5 nM for acetyl carnitine and 70 nM for carnitine. Relative standard deviations in a fivefold determination of standard solutions were between <2% for carnitine and <10% for trimethyllysine. Quantifying the formation of deuterated carnitine from deuterated gamma-butyrobetaine, this method is also suitable for the determination of the activity of gamma-butyrobetaine dioxygenase in tissues.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Nε,Nε,Nε-Trimethyllysine hydrochloride, ≥97% (TLC)