콘텐츠로 건너뛰기
Merck
  • 2-Phenyl and 2-heterocyclic-4-(3-(pyridin-2-yl)-1H-pyrazol-4-yl)pyridines as inhibitors of TGF-β1 and activin A signalling.
2-Phenyl and 2-heterocyclic-4-(3-(pyridin-2-yl)-1H-pyrazol-4-yl)pyridines as inhibitors of TGF-β1 and activin A signalling.
Bioorganic & medicinal chemistry letters (2011-07-26)
Rudy Ciayadi, Mahesh Potdar, Kelly L Walton, Craig A Harrison, Geoffrey F Kelso, Simon J Harris, Milton T W Hearn
초록
Novel inhibitors of TGF-β1 and activin A signalling based on a 2-aryl-4-(3-(pyridin-2-yl)-1H-pyrazol-4-yl)pyridine pharmacophore have been synthesised. Compounds containing phenyl or aromatic nitrogen heterocycle substituents inhibited both types of signalling with HEK-293T cells in culture, with a selectivity preference for TGF-β1. Synthetic compounds containing pyridin-3-yl, pyrazol-4-yl, pyrazol-1-yl or 1H-imidazoyl-1-yl substituents exhibited structural and functional attributes suitable for further investigation related to the development of more potent TGF-β inhibitors.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
1-Methylpyrazole-4-boronic acid pinacol ester, 95%