콘텐츠로 건너뛰기
Merck
  • Biochemical and genotoxic effects in women exposed to pesticides in Southern Ecuador.
Biochemical and genotoxic effects in women exposed to pesticides in Southern Ecuador.
Environmental science and pollution research international (2019-06-28)
Paulina Arévalo-Jaramillo, Alicia Idrobo, Lorena Salcedo, Andrea Cabrera, Andrea Vintimilla, Mayra Carrión, Natalia Bailon-Moscoso
초록
Toxicity refers to the potential of a substance such as a pesticide to cause damage to the structure or functions of an exposed organism. Pesticides can lead to harmful biological effects in exposed animals and their offspring over the medium and long term. They can affect the immunological, nervous, endocrine, and reproductive systems. DNA damage has also been linked to exposure to pesticides, and this damage can cause abortions, degenerative diseases, and cancer. The aim of this work was to establish whether women who are indirectly exposed to pesticides exhibit a compromised health status, including genotoxic effect. Women exposed indirectly to pesticides in Chimchanga and Colaisaca in the south of Ecuador underwent hematological and biochemical tests and micronucleus assay in buccal cells. The subjects were also genotyped for GSTM1, GSTT1, GSTP1, and PON1 polymorphisms, which can modify an individual's capacity to metabolize pesticides and relation with damage of DNA. The study revealed hepatic toxicity in Colaisaca women (AST and ALT) and an increase in the rate of micronucleus (MN) in Colaisaca individuals. In addition, genetic polymorphisms in PON1 and GSTP1 showed effects of modulating the frequency of karyolytic cells, karyorrhectic cells, and condensed chromatin cells.