콘텐츠로 건너뛰기
Merck
  • Virilizing sclerosing-stromal tumor of the ovary in a young woman with McCune Albright syndrome: clinical, pathological, and immunohistochemical studies.
Virilizing sclerosing-stromal tumor of the ovary in a young woman with McCune Albright syndrome: clinical, pathological, and immunohistochemical studies.
The Journal of clinical endocrinology and metabolism (2013-02-01)
Kahina Boussaïd, Geri Meduri, Jean-Christophe Maiza, Isabelle Gennero, Ghislaine Escourrou, Arnaud Bros, Pierre Leguevaque, Antoine Bennet, Philippe Caron
초록
McCune-Albright syndrome (MAS) is characterized by polyostotic fibrous dysplasia, café-au-lait skin pigmentations, and gonadotropin-independent sexual precocious puberty, resulting from a somatic postzygotic activating mutation of the GNAS1 gene. We report a virilizing sclerosing-stromal tumor of the ovary in a young female with MAS. She presented polyostotic fibrous dysplasia of the left upper and lower limbs and a café-au-lait skin spot in the posterior area of the neck. She had a history of precocious puberty, diagnosed at the age of 6 years and treated with cyproterone acetate until the age of 10 years; then she developed central puberty with severe oligomenorrhea. At the age of 23 years, she was hospitalized for a virilization syndrome including hirsutism, acne, deepening of the voice, amenorrhea, and clitoromegaly. Serum levels of T were dramatically increased (1293 ng/dl; normal range, 10-80). The abdominal computed tomography scan revealed a solid mass located on the left ovary. An ovariectomy was performed, and histological examination revealed a sclerosing-stromal tumor with pseudolobular pattern. Immunohistochemical studies revealed that the tumor cells expressed all steroidogenic enzymes involved in androgen synthesis. Molecular analysis revealed that ovarian tumor cells harbored the Arg 201 activating mutation in the GNAS1 gene. After surgery, T levels returned to normal, the patient retrieved a normal gonadal function, and she was able to become pregnant. This observation extends the clinical spectrum of ovarian pathology of women with MAS. However, the mechanisms causing this ovarian tumor remain unclear, even if the gsp oncogene has been implicated in the pathogenesis of some gonadal tumors.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
제품번호
팩 크기
재고 정보
가격
수량