콘텐츠로 건너뛰기
Merck
  • Capacitation-associated alkalization in human sperm is differentially controlled at the subcellular level.
Capacitation-associated alkalization in human sperm is differentially controlled at the subcellular level.
Journal of cell science (2020-01-15)
Arturo Matamoros-Volante, Claudia L Treviño
초록
Capacitation in mammalian sperm involves the accurate balance of intracellular pH (pHi), but the mechanisms controlling this process are not fully understood, particularly regarding the spatiotemporal regulation of the proteins involved in pHi modulation. Here, we employed an image-based flow cytometry technique combined with pharmacological approaches to study pHi dynamics at the subcellular level during capacitation. We found that, upon capacitation induction, sperm cells undergo intracellular alkalization in the head and principal piece regions. The observed localized pHi increases require the initial uptake of HCO3-, which is mediated by several proteins acting consistently with their subcellular localization. Hv1 proton channel (also known as HVCN1) and cAMP-activated protein kinase (protein kinase A, PKA) antagonists impair alkalization mainly in the principal piece. Na+/HCO3- cotransporter (NBC) and cystic fibrosis transmembrane regulator (CFTR) antagonists impair alkalization only mildly, predominantly in the head. Motility measurements indicate that inhibition of alkalization in the principal piece prevents the development of hyperactivated motility. Altogether, our findings shed light on the complex control mechanisms of pHi and underscore their importance during human sperm capacitation.This article has an associated First Person interview with the first author of the paper.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
KH7, ≥98% (HPLC)
Sigma-Aldrich
Propanoic acid hydrazide, ≥90% (GC)