콘텐츠로 건너뛰기
Merck
  • Photopolymerization of mixed monolayers and black lipid membranes containing gramicidin A and diacetylenic phospholipids.
Photopolymerization of mixed monolayers and black lipid membranes containing gramicidin A and diacetylenic phospholipids.
Langmuir : the ACS journal of surfaces and colloids (2006-01-25)
Susan M Daly, Linda A Heffernan, William R Barger, Devanand K Shenoy
초록
We formed monolayers and black lipid membranes (BLMs) of photopolymerizable lipids mixed with the channel-forming protein gramicidin A to evaluate their miscibility and the potential for improved stability of the BLM scaffold through polymerization. Analyses of surface pressure vs area isotherms indicated that gramicidin A dispersed with three different synthetic, polymerizable, diacetylene-containing phospholipids, 1,2-di-10,12-tricosadiynoyl-sn-glycero-3-phosphocholine (DTPC), 1,2-di-10,12-tricosadiynoyl-sn-glycero-3-phosphoethanolamine (DTPE), and 1-palmitoyl-2,10,12-tricosadiynoyl-sn-glycero-3-phosphoethanolamine (PTPE) to form mixed monolayers at the air-water interface on a Langmuir-Blodgett (LB) trough. Conductance measurements across a diacetylenic lipid-containing BLM confirmed dispersion of the gramicidin channel with the lipid layer and demonstrated gramicidin ion-channel activity before and after UV exposure. Polymerization kinetics of the diacetylenic films were monitored by film pressure changes at constant LB trough area and by UV-vis absorption spectroscopy of polymerized monolayers deposited onto quartz. An initial increase in film pressure of both the pure diacetylene lipid monolayers and mixed films upon exposure to UV light indicated a change in the film structure. Over the time scale of the pressure increase, an absorbance peak indicative of polymerization evolved, suggesting that the structural change in the lipid monolayer was due to polymerization. Film pressure and absorbance kinetics also revealed degradation of the polymerized chains at long exposure times, indicating an optimum time of UV irradiation for maximized polymerization in the lipid layer. Accordingly, exposure of polymerizable lipid-containing black lipid membranes to short increments of UV light led to an increase in the bilayer lifetime.
MATERIALS
제품 번호
브랜드
제품 설명
Avanti
23:2 Diyne PE [DC(8,9)PE], 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine, powder
Avanti
16:0-23:2 Diyne PE, 1-palmitoyl-2-(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine, powder