콘텐츠로 건너뛰기
Merck
  • Transition-Metal-Free Reductive Hydroxymethylation of Isoquinolines.
Transition-Metal-Free Reductive Hydroxymethylation of Isoquinolines.
Angewandte Chemie (International ed. in English) (2019-09-06)
Benjamin M Reeves, Hamish B Hepburn, Alexandru Grozavu, Peter J Lindsay-Scott, Timothy J Donohoe
초록
A transition-metal-free reductive hydroxymethylation reaction has been developed, enabling the preparation of tetrahydroisoquinolines bearing C4-quaternary centers from the corresponding isoquinolines. Deuterium labelling studies and control experiments enable a potential mechanism to be elucidated which features a key Cannizzaro-type reduction followed by an Evans-Tishchenko reaction. When isoquinolines featuring a proton at the 4-position are used, a tandem methylation-hydroxymethylation occurs, leading to the formation of 2 new C-C bonds in one pot.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
(2-Methylpropyl)boronic acid, ≥95.0%