콘텐츠로 건너뛰기
Merck
  • Girdin is a component of the lateral polarity protein network restricting cell dissemination.
Girdin is a component of the lateral polarity protein network restricting cell dissemination.
PLoS genetics (2020-03-21)
Cornélia Biehler, Li-Ting Wang, Myriam Sévigny, Alexandra Jetté, Clémence L Gamblin, Rachel Catterall, Elise Houssin, Luke McCaffrey, Patrick Laprise
초록
Epithelial cell polarity defects support cancer progression. It is thus crucial to decipher the functional interactions within the polarity protein network. Here we show that Drosophila Girdin and its human ortholog (GIRDIN) sustain the function of crucial lateral polarity proteins by inhibiting the apical kinase aPKC. Loss of GIRDIN expression is also associated with overgrowth of disorganized cell cysts. Moreover, we observed cell dissemination from GIRDIN knockdown cysts and tumorspheres, thereby showing that GIRDIN supports the cohesion of multicellular epithelial structures. Consistent with these observations, alteration of GIRDIN expression is associated with poor overall survival in subtypes of breast and lung cancers. Overall, we discovered a core mechanism contributing to epithelial cell polarization from flies to humans. Our data also indicate that GIRDIN has the potential to impair the progression of epithelial cancers by preserving cell polarity and restricting cell dissemination.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Anti-Girdin, coiled-coil region Antibody, from rabbit, purified by affinity chromatography