콘텐츠로 건너뛰기
Merck
  • Metabotropic signaling cascade involved in α4β2 nicotinic acetylcholine receptor-mediated PKCβII activation.
Metabotropic signaling cascade involved in α4β2 nicotinic acetylcholine receptor-mediated PKCβII activation.
Biochimica et biophysica acta. Molecular cell research (2020-04-19)
Srijan Acharya, Dooti Kundu, Hyun Jin Choi, Kyeong-Man Kim
초록
Nicotinic acetylcholine receptors (nAChRs) belong to the ionophore receptor family, which regulates plasma membrane conductance to Na+, K+, and Ca2+ ions. Some studies, however, have shown that nAChRs also employ second messengers for intracellular signaling. We previously showed that α4β2 nAChR mediates the translocation of protein kinase CβII (PKCβII) from the cytoplasm to the plasma membrane, which is a typical activation marker for PKCβII. In this study, we investigated the molecular mechanisms underlying PKCβII activation through α4β2 nAChR. α4β2 nAChR is the most abundant nAChR subtype and is implicated in various brain functions and diseases. Putative α4β2 nAChR signaling components were identified by knockdown or chemical inhibition of candidate proteins, and the signaling cascade was deduced by protein interactions in predicted cellular components. α4β2 nAChR-mediated PKCβII translocation was found to occur in an ionophore activity-independent manner. Nicotinic stimulation of α4β2 nAChR activated Src in a β-arrestin1 and 14-3-3η-dependent manner. Activated Src phosphorylated the tyrosine residue(s) on Syk molecules, which in turn interacted with phospholipase C γ1 to trigger the translocation of PKCβII to the cell membrane by elevating cellular diacylglycerol levels. The activated PKCβII in turn exerted a positive feedback effect on Src activation, suggesting that α4β2 nAChR signaling is amplified by a positive feedback loop. These findings provide novel information for unveiling the previously unclear metabotropic second messenger-based signal transduction pathway of nAChRs.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
BV02, ≥98% (HPLC)