콘텐츠로 건너뛰기
Merck
  • Efficiency of Modified Triple-Branched Stent Graft in Type I Aortic Dissection: Two-Year Follow-up.
Efficiency of Modified Triple-Branched Stent Graft in Type I Aortic Dissection: Two-Year Follow-up.
The Annals of thoracic surgery (2020-01-29)
Zhi-Huang Qiu, Liang-Wan Chen, Lian-Ming Liao, Jun Xiao, Xiao-Fu Dai, Guan-Hua Fang, Liang-Liang Yan, Qing-Song Wu, Quan-Fang Luo
초록
The efficacy of hemiarch replacement combined with a modified triple-branched stent graft in Debakey type I aortic dissection remains to be confirmed. From January 2016 to December 2017, 167 patients with acute Debakey type I aortic dissection underwent hemiarch replacement combined with a modified triple-branched stent graft. The clinical and imaging data were retrospectively analyzed. The early composite endpoint was defined to comprise perioperative mortality, permanent neurologic deficits, and renal failure requiring hemodialysis at discharge. The overall 30-day mortality was 4.2% (7 of 167). The incidence of the composite endpoint was 11.4% (19 of 167). The risk factors for the composite endpoint were malperfusion syndrome (odds ratio 5.17; 95% confidence interval, 1.46 to 18.35; P = .011) and creatine greater than 1.5 mg/dL (odds ratio 5.44; 95% confidence interval, 2.27 to 13.06; P < .001). The overall survival was 94% at 1 year and 92.2% at 2 years during a median follow-up of 20.9 ± 9.6 months. Three patients required distal aorta reintervention. Complete thrombosis in the false lumen of the descending aorta at the level of the pulmonary bifurcation and at the level of the celiac trunk was observed in 98.8% and 10.8% of the patients, respectively. Hemiarch replacement combined with a modified triple-branched stent graft is a reliable technique for acute Debakey type I aortic dissection as indicated by 2 years of follow-up.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Diisopropyl azodicarboxylate, 98%