콘텐츠로 건너뛰기
Merck
  • Tungsten-niobium oxide bronzes: a bulk and surface structural study.
Tungsten-niobium oxide bronzes: a bulk and surface structural study.
Dalton transactions (Cambridge, England : 2003) (2020-09-17)
Daniel Delgado, Patricia Concepción, Annette Trunschke, Jose M López Nieto
초록
Materials from the WO3-Nb2O5 system, presenting bronze-type crystal structures, display outstanding functional properties for several applications as thermoelectric materials, lithium-ion battery electrodes, or catalysts. In this work, a series of W-Nb-O oxide bronzes have been synthesized by the hydrothermal method (with Nb/(W + Nb) ratios in the range of 0-1). A combination of bulk and surface characterisation techniques has been applied to get further insights into: (i) the effect of thermal treatments on as-prepared materials and (ii) the surface chemical nature of W-Nb-O oxide bronzes. Thermal treatments promote the following structural changes: (i) loss of emerging long-range order and (ii) the elimination of NH4+ and H2O species from the structural channels of the as-synthesized materials. It has been observed that W-Nb-O bronzes with Nb at% of ca. 50% are able to retain a long-range order after heat-treatments, which is attributed to the presence of a Cs0.5[W2.5Nb2.5O14]-type structure. Increasing amounts of Nb5+ in the materials (i) promote a phase transition to pseudocrystalline phases ordered along the c-axis; (ii) stabilize surface W5+ species (elucidated by XPS); and (iii) increase the proportion of surface Lewis acid sites (as determined by the FTIR of adsorbed CO). Results suggest that pseudocrystalline oxides (with a Nb at% ≥ 50%) are closely related to NbO7 pentagonal bipyramid-containing structures. The stabilisation of Lewis acid sites on these pseudocrystalline materials leads to a higher yield of heavy compounds, at the expense of acrolein formation, in the gas-phase dehydration of glycerol.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Ammonium metatungstate hydrate, ≥85% WO3 basis (gravimetric)