콘텐츠로 건너뛰기
Merck
Curcumin is a potent DNA hypomethylation agent.
Bioorganic & medicinal chemistry letters (2008-12-30)
Zhongfa Liu, Zhiliang Xie, William Jones, Ryan E Pavlovicz, Shujun Liu, Jianhua Yu, Pui-kai Li, Jiayuh Lin, Jame R Fuchs, Guido Marcucci, Chenglong Li, Kenneth K Chan
초록
Molecular docking of the interaction of curcumin and DNMT1 suggested that curcumin covalently blocks the catalytic thiolate of C1226 of DNMT1 to exert its inhibitory effect. This was validated by showing that curcumin inhibits the activity of M. SssI with an IC(50) of 30 nM, but no inhibitory activity of hexahydrocurcumin up to 100 microM. In addition, curcumin can induce global DNA hypomethylation in a leukemia cell line.
MATERIALS
제품 번호
브랜드
제품 설명
Supelco
Bisdemethoxycurcumin, analytical standard