콘텐츠로 건너뛰기
Merck
  • Combination of Self-Healing Butyl Rubber and Natural Rubber Composites for Improving the Stability.
Combination of Self-Healing Butyl Rubber and Natural Rubber Composites for Improving the Stability.
Polymers (2021-02-13)
Kunakorn Chumnum, Ekwipoo Kalkornsurapranee, Jobish Johns, Karnda Sengloyluan, Yeampon Nakaramontri
초록
The self-healing composites were prepared from the combination of bromobutyl rubber (BIIR) and natural rubber (NR) blends filled with carbon nanotubes (CNT) and carbon black (CB). To reach the optimized self-healing propagation, the BIIR was modified with ionic liquid (IL) and butylimidazole (IM), and blended with NR using the ratios of 70:30 and 80:20 BIIR:NR. Physical and chemical modifications were confirmed from the mixing torque and attenuated total reflection-fourier transform infrared spectroscopy (ATR-FTIR). It was found that the BIIR/NR-CNTCB with IL and IM effectively improved the cure properties with enhanced tensile properties relative to pure BIIR/NR blends. For the healed composites, BIIR/NR-CNTCB-IM exhibited superior mechanical and electrical properties due to the existing ionic linkages in rubber matrix. For the abrasion resistances, puncture stress and electrical recyclability were examined to know the possibility of inner liner applications and Taber abrasion with dynamic mechanical properties were elucidated for tire tread applications. Based on the obtained Tg and Tan δ values, the composites are proposed for tire applications in the future with a simplified preparation procedure.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
1-Butylimidazole, 98%