콘텐츠로 건너뛰기
Merck
5-benzyloxytryptamine as an antagonist of TRPM8.
Bioorganic & medicinal chemistry letters (2010-10-23)
Jeff DeFalco, Daniel Steiger, Michelle Dourado, Daniel Emerling, Matthew A J Duncton
초록
5-Benzyloxytryptamine 19 was found to act as an antagonist of the TRPM8 ion-channel. For example, 19 had an IC(50) of 0.34 μM when menthol was used as the stimulating agonist. Related commercially-available tryptamine derivatives showed diminished, or no antagonist activity at TRPM8. The structural similarity of 5-benzyloxytryptamine to other literature TRPM8 antagonists was noted.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
5-Methoxytryptamine, 97%
Sigma-Aldrich
5-(Benzyloxy)indole, 95%