콘텐츠로 건너뛰기
Merck
  • Transparency Change Mechanochromism Based on a Robust PDMS-Hydrogel Bilayer Structure.
Transparency Change Mechanochromism Based on a Robust PDMS-Hydrogel Bilayer Structure.
Macromolecular rapid communications (2020-10-28)
Yonghang Xu, Songshan Zeng, Weikang Xian, Limiao Lin, Hao Ding, Jingjing Liu, Min Xiao, Shuanjin Wang, Ying Li, Yuezhong Meng, Luyi Sun
초록
Hydrogels and polydimethylsiloxane (PDMS) are complementary to each other, since the hydrophobic PDMS provides a more stable and rigid substrate, while the water-rich hydrogel possesses remarkable hydrophilicity, biocompatibility, and similarity to biological tissues. Herein a transparent and stretchable covalently bonded PDMS-hydrogel bilayer (PHB) structure is prepared via in situ free radical copolymerization of acrylamide and allylamine-exfoliated-ZrP (AA-e-ZrP) on a functionalized PDMS surface. The AA-e-ZrP serves as cross-linking nano-patches in the polymer gel network. The covalently bonded structure is constructed through the addition reaction of vinyl groups of PDMS surface and monomers, obtaining a strong interfacial adhesion between the PDMS and the hydrogel. A mechanical-responsive wrinkle surface, which exhibs transparency change mechanochromism, is created via introducing a cross-linked polyvinyl alcohol film atop the PHB structure. A finite element model is implemented to simulate the wrinkle formation process. The implication of the present finding for the interfacial design of the PHB and PDMS-hydrogel-PVA trilayer (PHPT) structures is discussed.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Allylamine, ≥99%