콘텐츠로 건너뛰기
Merck
  • Inhibition and enhancement of odorant-induced cAMP accumulation in rat olfactory cilia by antibodies directed against G alpha S/olf- and G alpha i-protein subunits.
Inhibition and enhancement of odorant-induced cAMP accumulation in rat olfactory cilia by antibodies directed against G alpha S/olf- and G alpha i-protein subunits.
FEBS letters (1998-05-26)
S Sinnarajah, P I Ezeh, S Pathirana, A G Moss, E E Morrison, V Vodyanoy
초록
The odorant-induced accumulation of cAMP can be inhibited by antibodies directed against G alpha s/olf. In contrast, antibodies raised against G alpha i-subunits caused a strong enhancement of the odorant-induced cAMP accumulation. Western blotting and immunoelectron microscopy revealed the presence of both G alpha s/olf- and G alpha i-subunits in rat cilia preparations. The existence of both stimulatory and inhibitory odorant-induced regulation of adenylyl cyclase activity in olfactory cilia may indicate that an initial integration of different odorant stimuli begins at the level of primary reactions in the same effector enzyme.