콘텐츠로 건너뛰기
Merck
  • CDK5 inhibition improves glucose uptake in insulin-resistant neuronal cells via ERK1/2 pathway.
CDK5 inhibition improves glucose uptake in insulin-resistant neuronal cells via ERK1/2 pathway.
Cell biology international (2021-12-06)
Kapil Manglani, Chinmoy S Dey
초록
Role of CDK5 and its inhibition in various neuronal processes and functions are well established. However, role of CDK5 and its inhibition in neuronal insulin-signaling and-resistance is not yet explored. In the present study, we investigated the effect of CDK5 inhibition in neuronal insulin signaling, specifically insulin-stimulated glucose uptake. CDK5 expression in neuro-2a cells was increased under insulin-resistant state, developed by chronic treatment of insulin, confirming the crucial role of CDK5 in insulin resistance in neuronal cells. However, whether increased expression of CDK5 in hyperinsulinemia-mediated insulin-resistant conditions is a cause or a consequence, is still an unanswered question. We showed that CDK5 inhibition did not affect basal insulin signaling; however, insulin-stimulated glucose uptake enhanced in insulin-resistant cells. Moreover, CDK5 inhibition could improve glucose uptake, the ultimate outcome of insulin signaling, in insulin-resistant neuro-2a cells. We first time showed that CDK5 inhibition by roscovitine could ameliorate insulin resistance and increase glucose uptake in neuronal cells via ERK1/2 pathway. Our study provides intriguing insights about the effect of CDK5 inhibition on neuronal insulin resistance and opens up a new paradigm to develop new therapeutic strategies for neuronal insulin resistance and associated pathophysiological conditions.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Insulin, Zinc, Human, Recombinant, P. pastoris
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution