콘텐츠로 건너뛰기
Merck
  • Anti-influenza drug discovery: structure-activity relationship and mechanistic insight into novel angelicin derivatives.
Anti-influenza drug discovery: structure-activity relationship and mechanistic insight into novel angelicin derivatives.
Journal of medicinal chemistry (2010-01-23)
Jiann-Yih Yeh, Mohane Selvaraj Coumar, Jim-Tong Horng, Hui-Yi Shiao, Fu-Ming Kuo, Hui-Ling Lee, In-Chun Chen, Chun-Wei Chang, Wen-Fang Tang, Sung-Nain Tseng, Chi-Jene Chen, Shin-Ru Shih, John T-A Hsu, Chun-Chen Liao, Yu-Sheng Chao, Hsing-Pang Hsieh
초록
By using a cell-based high throughput screening campaign, a novel angelicin derivative 6a was identified to inhibit influenza A (H1N1) virus induced cytopathic effect in Madin-Darby canine kidney cell culture in low micromolar range. Detailed structure-activity relationship studies of 6a revealed that the angelicin scaffold is essential for activity in pharmacophore B, while meta-substituted phenyl/2-thiophene rings are optimal in pharmacophore A and C. The optimized lead 4-methyl-9-phenyl-8-(thiophene-2-carbonyl)-furo[2,3-h]chromen-2-one (8g, IC(50) = 70 nM) showed 64-fold enhanced activity compared to the high throughput screening (HTS) hit 6a. Also, 8g was found effective in case of influenza A (H3N2) and influenza B virus strains similar to approved anti-influenza drug zanamivir (4). Preliminary mechanistic studies suggest that these compounds act as anti-influenza agents by inhibiting ribonucleoprotein (RNP) complex associated activity and have the potential to be developed further, which could form the basis for developing additional defense against influenza pandemics.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Angelicin