콘텐츠로 건너뛰기
Merck
  • Acid-triggered transformation of diortho ester phosphocholine liposome.
Acid-triggered transformation of diortho ester phosphocholine liposome.
Journal of the American Chemical Society (2006-01-05)
Zhaohua Huang, Xin Guo, Weijun Li, J Andrew MacKay, Francis C Szoka
초록
The use of pH-sensitive liposomes for acid-triggered site-specific drug delivery is one of the more promising approaches to improve the therapeutic index of drugs. Here, we report the synthesis, assembly, and hydrolysis of the first ortho ester phosphocholine (OEPC). The acid hydrolysis of OEPC liposomes consists of a lag phase and a burst phase. The lag time is pH-dependent-the lower the pH, the shorter the time. Upon acid hydrolysis, the OEPC liposomes were transformed into leaky metastable vesicles that rapidly collapsed in the presence of albumin. OEPC, when formulated with cationic lipid, significantly enhanced the transfection efficiency compared with that of the pH-insensitive formulation.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Oleyl alcohol, ≥99% (GC)
Sigma-Aldrich
Oleyl alcohol, technical grade, 85%