콘텐츠로 건너뛰기
Merck
  • [Progress in studies of pharmacological action and mechanisms of catalpol on brain diease].
[Progress in studies of pharmacological action and mechanisms of catalpol on brain diease].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2010-03-13)
Huifeng Zhu, Dong Wan, Fen Zhang
초록
Catalpol is a effective components of rehmannia root, it have many pharmacological actions, such as anti-brain ischemia, anti-senile dementia, promoting neuro-remodeling and reducing capillary permeability and so on.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Catalpol, ≥96% (HPLC)