콘텐츠로 건너뛰기
Merck
  • Fatal cardiac arrhythmia after repeated exposure to 1,1-difluoroethane (DFE).
Fatal cardiac arrhythmia after repeated exposure to 1,1-difluoroethane (DFE).
The American journal of forensic medicine and pathology (2006-02-28)
Joseph Avella, James C Wilson, Michael Lehrer
초록
A 42-year-old man was found dead after repeated exposure to 1,1-difluoroethane (DFE, Freon 152a), a propellant found in CRC Duster, a product intended for the removal of dust and lint. Toxicologic analysis detected DFE in femoral blood 136.3 mg/L, brain 117.5 mg/kg, liver 87.6 mg/kg, lung 60.3 mg/kg, adipose 235.7 mg/kg, and vitreous fluid 25.1 mg/L. The cause of death was determined to be a fatal cardiac arrhythmia due to intoxication with 1,1-difluoroethane. After comparison to previously published cases involving DFE, we suggest that analysis of adipose tissue for DFE and similar compounds, along with blood and other tissues, may be useful in distinguishing between acute versus chronic exposure. Adipose may also be a valuable alternate specimen for detection in cases where loss or elimination from blood is likely to have occurred.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
1,1-Difluoroethane, ≥98%