콘텐츠로 건너뛰기
Merck
  • Substituted 2-phenyl-benzimidazole derivatives: novel compounds that suppress key markers of allergy.
Substituted 2-phenyl-benzimidazole derivatives: novel compounds that suppress key markers of allergy.
European journal of medicinal chemistry (2006-05-02)
Mark L Richards, Shirley Cruz Lio, Anjana Sinha, Homayon Banie, Richard J Thomas, Michael Major, Mark Tanji, Jagadish C Sircar
초록
The pharmacotherapy of allergy and asthma has traditionally focused on the effecter molecules of the allergic cascade, while neglecting targets that play an early role in their development. Reasoning that IgE is central to the expansion of atopic diseases, we identified and extended a novel family of 2-(substituted phenyl)-benzimidazole inhibitors of IgE response. Pharmacological activity depends on an intact phenylbenzimidazole-bis-amide backbone, and is optimized by the presence of lipophilic terminal groups composed of either bis cycloalkyl or combinations of aliphatic and halogen-substituted aromatic groups. These compounds also inhibit IL-4 and IL-5 responses in T cells and CD23 expression on B cells, with potencies that parallel their inhibition of IgE. The broad profile of these compounds thus underscores their potential for treating the multifarious pathology of asthma.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
2-Phenylbenzimidazole, 97%