콘텐츠로 건너뛰기
Merck
  • A case of an accidental exposure to a veterinary insecticide product formulation.
A case of an accidental exposure to a veterinary insecticide product formulation.
Veterinary and human toxicology (1989-02-01)
K S Sidhu, M B Collisi
초록
A veterinary technician while opening a package was accidentally exposed to a commercial canned product formulation containing insecticides and solvents. The patient was twice briefly treated and released as an outpatient from 2 different hospitals on the first and second day after the exposure. However, on the fourth day, as some of the symptoms (headache, nausea, vomiting, diarrhea, difficult breathing) persisted, the patient was admitted to another hospital. The patient was treated for exposure to organophosphates and solvents and was released after 13 days. The patient developed diabetes insipidus, a condition which lasted for approximately 1 year. The cause of the temporary development of diabetes insipidus is not understood. There is a need to prevent and minimize such accidental exposures in future.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
1,1-Difluoroethane, ≥98%